Ethyl-α-bromophenylacetate 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Ethyl alpha-bromophenylacetate
438060050
438060250
438061000
ACRO438060050EA
1300
CZK
ACRO438060050
ACRO438060250
ACRO438061000
Ethyl-α-bromophenylacetate 97%
Ethyl-α-bromophenylacetate
Vzorec:
C₁₀H₁₁BrO₂ MW: 243.10 g/mol Bod varu: 89…92 °C (1.2hPa) Hustota: 1.389 g/cm³ Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013536 CAS číslo: 2882-19-1 UN: 3265 ADR: 8 II |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5370 to 1.5420 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...