Cyclopentanesulphonyl chloride 90%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Cyclopentanesulphonylchloride
437830010
437830050
ACRO437830010EA
5230
CZK
ACRO437830010
ACRO437830050
Cyclopentanesulphonyl chloride 90%
Cyclopentanesulphonyl chloride
Vzorec:
C₅H₉ClO₂S Bod varu: 95…98 °C (2Torr) Bod vzplanutí: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04117774 CAS číslo: 26394-17-2 ADR: 8,II |
Výsledky specifikačního testu
Appearance | Light brown to dark brown Liquid |
Infrared spectrum | Conforms |
GC | ≥88.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...