1-Boc-4-piperidineacetic acid 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
[1-(tert-Butoxycarbonyl)piperidin-4-yl]acetic acid, 2-(1-(tert-Butoxycarbonyl)piperidin-4-yl)acetic acid, 1-(tert-Butoxycarbonyl)-4-piperidylacetic acid
437500010
437500050
ACRO437500010EA
5700
CZK
ACRO437500010
ACRO437500050
1-Boc-4-piperidineacetic acid 97%
1-Boc-4-piperidineacetic acid
Vzorec:
C₁₂H₂₁NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00800239 CAS číslo: 157688-46-5 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 96 to 102 °C |
Titration with NaOH | 96 to 104 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...