Tisk…

(S)-(+)-3-Amino-1-boc-piperidine 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (S)-1-Boc-3-aminopiperidine, (S)-tert-Butyl 3-aminopiperidine-1-carboxylate, (S)-(+)-3-Amino-1-BOC-piperidine

437490010 437490050
ACRO437490010EA 5450 CZK
ACRO437490010 ACRO437490050
(S)-(+)-3-Amino-1-boc-piperidine 95%
(S)-(+)-3-Amino-1-boc-piperidine
Vzorec: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094718
CAS číslo: 625471-18-3
UN: 2735
ADR: 8 III

Objednat

Výsledky specifikačního testu

Appearance Colorless to yellow Gum or semi-solid
Infrared spectrum Conforms
GC ≥94.0 %
Specific optical rotation +26.5° to +30.5° (20°C, 589 nm) (c=1 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR