Tisk…

(S)-3-(Boc-amino)piperidine 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (S)-3-Boc-aminopiperidine, (S)-tert-Butyl piperidin-3-ylcarbamate, tert-butyl N-[(3S)-piperidin-3-yl]carbamate

436530010 436530050
ACRO436530010EA 2050 CZK
ACRO436530010 ACRO436530050
(S)-3-(Boc-amino)piperidine 97%
(S)-3-(Boc-amino)piperidine
Vzorec: C₁₀H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03093383
CAS číslo: 216854-23-8

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation -2.5° to -3.5° (20°C, 589 nm) (c=1 in DMF)
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR