2-Fluoro-4-pyridineboronic acid pinacol ester 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Fluoropyridine-4-boronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
435930010
435930050
ACRO435930010EA
5230
CZK
ACRO435930010
ACRO435930050
2-Fluoro-4-pyridineboronic acid pinacol ester 95%
2-Fluoro-4-pyridineboronic acid pinacol ester
Vzorec:
C₁₁H₁₅BFNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD06798253 CAS číslo: 458532-86-0 |
Výsledky specifikačního testu
Appearance | Yellow to beige Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 47 to 54 °C |
GC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...