Tisk…

tert-Butyl-N-allylcarbamate 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: N-Boc-allylamine, N-(tert-Butoxycarbonyl)allylamine, N-Allylcarbamic Acid tert-Butyl Ester, tert-Butyl allylcarbamate

434770100 434770500
ACRO434770100EA 2220 CZK
ACRO434770100 ACRO434770500
tert-Butyl-N-allylcarbamate 98%
tert-Butyl-N-allylcarbamate
Vzorec: C₈H₁₅NO₂
Bod varu: 48…50 °C (1 mmHg)
Bod vzplanutí: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00191870
CAS číslo: 78888-18-3

Objednat

Výsledky specifikačního testu

Appearance Colorless to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 33 to 40 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR