Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
methyl 4-oxotetrahydro-2H-thiopyran-3-carboxylate, methyl-4-oxotetrahydro-2H-thiopyran-3-carboxylate, Methyl 4-oxothiane-3-carboxylate
434290010
434290050
ACRO434290010EA
5190
CZK
ACRO434290010
ACRO434290050
Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate 95%
Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate
Vzorec:
C₇H₁₀O₃S MW: 174,214 g/mol Bod varu: 134 °C (10Torr) Storage Temperature: Ambient |
MDL Number:
MFCD00203485 CAS číslo: 4160-61-6 |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid or solid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...