Vanadium(III) chloride tetrahydrofuran complex (1:3) 0.5M in methylene chloride, AcroSeal®
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
433661000
433668000
433661000.EA
3470
CZK
433661000.
433668000.
Vanadium(III) chloride tetrahydrofuran complex (1:3) 0.5M in methylene chloride, AcroSeal®
Vanadium(III) chloride tetrahydrofuran complex (1:3)
Vzorec:
VCl₃·3C₄H₈O Hustota: 1,312 g/cm³ Bod vzplanutí: –8 °C Storage Temperature: Ambient |
MDL Number:
MFCD29050135 CAS číslo: 19559-06-9 UN: 1593 ADR: 6.1,III |
Výsledky specifikačního testu
Appearance | Light red to dark red Solution |
Titration Argentometric | 0.40 to 0.60 M |
Water | ≤0.005 % |
GC | ≥99.4 % |
Stabilizer | Approx. 50 ppm BHT |
Peroxide value | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...