Tisk…

3-Aminomethyltetrahydrofuran 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

433000010 433000050
ACRO433000010EA 8610 CZK
ACRO433000010 ACRO433000050
3-Aminomethyltetrahydrofuran 97%
3-Aminomethyltetrahydrofuran
Vzorec: C₅H₁₁NO
Bod varu: 65 °C (15Torr)
Storage Temperature: Ambient
MDL Number: MFCD08234925
CAS číslo: 165253-31-6
UN: 2924
ADR: 3,III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Residue after ignition ≤0.1 %
pH 9.5 to 11.0 (5% at 25°C)
Titration with HClO4 ≥99 %
Iron (Fe) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR