Tisk…

Methyl-6-chloropyridazine-4-carboxylate 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Methyl 6-chloropyridazine-4-carboxylate

432980010 432980050
ACRO432980010EA 5960 CZK
ACRO432980010 ACRO432980050
Methyl-6-chloropyridazine-4-carboxylate 97%
Methyl-6-chloropyridazine-4-carboxylate
Vzorec: C₆H₅ClN₂O₂
MW: 172,5691 g/mol
Bod tání: 88,4…88.5 °C
Storage Temperature: Ambient
MDL Number: MFCD16660968
CAS číslo: 1093860-48-0

Objednat

Výsledky specifikačního testu

Appearance Cream to brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 81 to 89 °C
GC ≥96.0 %
UV at 230 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.18 M soln.) Passes test
Suitability (HPLC gradient) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR