Tisk…

4-Methoxybenzenesulphonamide 95%

Dodavatel: Thermo Scientific
Varování

432910050 432910250
ACRO432910050EA 2600 CZK
ACRO432910050 ACRO432910250
4-Methoxybenzenesulphonamide 95%
4-Methoxybenzenesulphonamide
Vzorec: C₇H₉NO₃S
Bod tání: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00025392
CAS číslo: 1129-26-6

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 109 to 113 °C
HPLC ≥94.0 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR