Tisk…

[(tert-Butoxycarbonyl)aminooxy]acetic acid 98%

Dodavatel: Thermo Fisher Scientific
Varování

429690010 429690050
ACRO429690010EA 2080 CZK
ACRO429690010 ACRO429690050
[(tert-Butoxycarbonyl)aminooxy]acetic acid 98%
[(tert-Butoxycarbonyl)aminooxy]acetic acid
Vzorec: C₇H₁₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD01632027
CAS číslo: 42989-85-5

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 113 °C (Dec)
Titration with HClO4 ≥97.5 %
Loss on ignition 24.5 % to 26.5 % (800 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR