Benzyl-4-oxo-1-piperidinecarboxylate 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Cbz-Piperidin-4-one, 1-Carbobenzoxy-4-piperidone, 1-Cbz-4-piperidone, 4-Oxo-1-piperidinecarboxylic Acid Benzyl Ester, 1-Benzyloxycarbonyl-4-piperidine
425960050
425960250
ACRO425960050EA
1130
CZK
ACRO425960050
ACRO425960250
Benzyl-4-oxo-1-piperidinecarboxylate 97%
Benzyl-4-oxo-1-piperidinecarboxylate
Vzorec:
C₁₃H₁₅NO₃ MW: 233,27 g/mol Bod varu: 114…140 °C (,3 mmHg) Hustota: 1,185 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00673144 CAS číslo: 19099-93-5 |
Výsledky specifikačního testu
Appearance | Colorless to white to light yellow to light orange Powder or solid or liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...