Corn oil
Dodavatel: Thermo Fisher Scientific
405430025
405435000
ACRO405430025EA
5960
CZK
ACRO405430025
ACRO405435000
Corn oil
Corn oil
Bod tání:
-10 °C Hustota: 0,92 g/cm³ Bod vzplanutí: >200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130865 CAS číslo: 8001-30-7 |
Výsledky specifikačního testu
Appearance | Clear yellow Oily liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4730 to 1.4760 (20°C, 589 nm) |
Acid value | ≤0.5 mg KOH/g |
Peroxide value | ≤5 meq O2/kg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...