Methyl hydrogen isophthalate 97%
Dodavatel: Thermo Scientific
Synonyms:
Isophthalic acid monomethyl ester, 3-(Methoxycarbonyl)benzoic acid, Benzene-1,3-dicarboxylic acid monomethyl ester, mono-Methyl isophthalate
398230010
398230050
ACRO398230010EA
1440
CZK
ACRO398230010
ACRO398230050
Methyl hydrogen isophthalate 97%
Methyl hydrogen isophthalate
Vzorec:
C₉H₈O₄ MW: 180,16 g/mol Bod tání: 194…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00029972 CAS číslo: 1877-71-0 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 191 to 197 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...