Tisk…

tert-Butyl (diethylphosphono)acetate 95%

Dodavatel: Thermo Scientific
Varování

Synonyms: tert-Butyl 2-(diethoxyphosphoryl)acetate, Diethyl(2-tert-butoxycarbonylmethyl)phosphonate

397750010 397750050
ACRO397750010EA 1090 CZK
ACRO397750010 ACRO397750050
tert-Butyl (diethylphosphono)acetate 95%
tert-Butyl (diethylphosphono)acetate
Vzorec: C₁₀H₂₁O₅P
MW: 252,25 g/mol
Hustota: 1,074 g/cm³
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075414
CAS číslo: 27784-76-5

Objednat

Výsledky specifikačního testu

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR