Tisk…

Methylthymol blue sodium salt ACS indicator grade

Dodavatel: Thermo Fisher Scientific

Synonyms: Sodium 2,2',2'',2'''-((((1,1-dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetate, Methylthymol Blue tetrasodium salt

394210010 394210050
ACRO394210010EA 1770 CZK
ACRO394210010 ACRO394210050
Methylthymol blue sodium salt ACS indicator grade
Methylthymol blue sodium salt
Vzorec: C₃₇H₄₀N₂Na₄O₁₃S
Storage Temperature: Ambient
MDL Number: MFCD00148905
CAS číslo: 1945-77-3

Objednat

Výsledky specifikačního testu

Appearance Dark green to brown to black Powder
Infrared spectrum Conforms
Visual transition interval From pH 6.5 (yellow) to pH 8.5 (blue)
Suitability (as metal indicator) Passes test
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR