Thioxanthene 98%
Dodavatel: Thermo Scientific
391670010
391670050
ACRO391670010EA
3070
CZK
ACRO391670010
ACRO391670050
Thioxanthene 98%
Thioxanthene
Bod varu:
340 °C (730 mmHg) Bod tání: 128…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046936 CAS číslo: 261-31-4 |
Výsledky specifikačního testu
Appearance | White Crystals or needles |
Infrared spectrum | Conforms |
Melting point | 127 to 131 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...