Rhodium(II) trifluoroacetate dimer
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Trifluoroacetic acid rhodium(II) salt dimer
389700010
389702500
ACRO389700010EA
19120
CZK
ACRO389700010
ACRO389702500
Rhodium(II) trifluoroacetate dimer
Rhodium(II) trifluoroacetate dimer
Vzorec:
C₈F₁₂O₈Rh₂ MW: 657.87 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00209611 CAS číslo: 31126-95-1 |
Výsledky specifikačního testu
Appearance | Green Crystalline powder |
Infrared spectrum | Conforms |
Rhodium (Rh) | 27.5 to 35.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...