Tisk…

Bis(trifluoromethanesulphonyl)imide 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: lithium bis(trifluoromethanesulphonimide), 1,1,1-Trifluoro-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N,N-Bis(trifluoromethylsulphonyl)amine

387600010 387600050 387600250
ACRO387600010EA 1800 CZK
ACRO387600010 ACRO387600050 ACRO387600250
Bis(trifluoromethanesulphonyl)imide 99%
Bis(trifluoromethanesulphonyl)imide
Vzorec: C₂HF₆NO₄S₂
MW: 281.143 g/mol
Bod varu: 90…91 °C (760 mmHg)
Bod tání: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00214154
CAS číslo: 82113-65-3
UN: 3261
ADR: 8 I

Objednat

Výsledky specifikačního testu

Appearance Colorless to brown Crystals
Infrared spectrum Conforms
Melting point 50 to 56 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR