Tisk…

Sodium octadecyl sulphate 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Sodium Stearyl sulphate, n-Octadecyl sulphate, sodium salt, Sodium n-octadecylsulphate, Octadecyl sulphuric acid salt

384890010 384890050 384890250
ACRO384890010EA 2160 CZK
ACRO384890010 ACRO384890050 ACRO384890250
Sodium octadecyl sulphate 98%
Sodium octadecyl sulphate
Vzorec: C₁₈H₃₇NaO₄S
Bod tání: 211…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00007469
CAS číslo: 1120-04-3

Objednat

Výsledky specifikačního testu

Appearance White to light yellow to light beige Powder
Infrared spectrum Conforms
Loss on drying ≤7 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR