Methyl-ɑ-bromophenylacetate 97+%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
379300050
379300250
ACRO379300050EA
1660
CZK
ACRO379300050
ACRO379300250
Methyl-ɑ-bromophenylacetate 97+%
Methyl-alpha-bromophenylacetate
Vzorec:
C₉H₉BrO₂ Bod varu: 113 °C (3 mmHg) Hustota: 1,455 g/cm³ Bod vzplanutí: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013535 CAS číslo: 3042-81-7 UN: 3265 ADR: 8,II |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...