Chlorobis(cyclooctene)rhodium(I) dimer 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
369371000
369375000
ACRO369371000EA
2160
CZK
ACRO369371000
ACRO369375000
Chlorobis(cyclooctene)rhodium(I) dimer 98%
Chlorobis(cyclooctene)rhodium(I) dimer
Vzorec:
C₃₂H₅₆Cl₂Rh₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00013287 CAS číslo: 12279-09-3 |
Výsledky specifikačního testu
Appearance | Orange-brown Powder |
Infrared spectrum | Conforms |
Rhodium (Rh) | 28.0 to 29.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...