2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide 95% stabilizovaný
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Acetobromo-α-D-galactose, (2R,3S,4S,5R,6R)-2-(acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, Tetra-O-acetyl-α-D-galactopyranosyl bromide, Acetobromo-a-D-galactose
353690100
353690500
ACRO353690100EA
5630
CZK
ACRO353690100
ACRO353690500
2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide 95% stabilizovaný
2,3,4,6-Tetra-O-acetyl-alpha-D-galactopyranosyl bromide
Vzorec:
C₁₄H₁₉BrO₉ MW: 411,2 g/mol Bod tání: 83…85 °C Storage Temperature: Freezer |
MDL Number:
MFCD00063686 CAS číslo: 3068-32-4 |
Výsledky specifikačního testu
Appearance | White to light beige Powder or crystals and/or chunks (May darken on storage) |
Infrared spectrum | Conforms |
Titration Argentometric | ≥94 % |
Stabilizer | 2 % CaCO3 |
Specific optical rotation | +200° to +220° (20°C, 589 nm) (c=3, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...