Tisk…

Vitamin A acetate 1,5 MIU/g in sunflower oil stabilizovaný

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: all-trans-Retinyl acetate, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate

346321000 346325000
ACRO346321000EA 4780 CZK
ACRO346321000 ACRO346325000
Vitamin A acetate 1,5 MIU/g in sunflower oil stabilizovaný
Vitamin A acetate
Vzorec: C₂₂H₃₂O₂
MW: 328,49 g/mol
Bod vzplanutí: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00019413
CAS číslo: 127-47-9
UN: 3082
ADR: 9,III

Objednat

Výsledky specifikačního testu

Appearance Yellow to brown-yellow Partly crystalline when cooled
Assay 1.50 to 1.65 million IU/g (Vitamin A)
Identification (TLC): conforms
Stabilizer BHT
Related substances ≤0.14 (A370/A326)
Acid value ≤2.0 mg KOH/g
Peroxide value ≤15 meq/kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR