Tisk…

tert-Butyl (3R)-3-amino-3-phenylpropanoate 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1,1-dimethylethyl (3R)-3-amino-3-phenylpropanoate, (3R)-3-Phenyl-beta-alanine tert-butyl ester, Benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester,(bR)-

337882500 337880010
ACRO337882500EA 5780 CZK
ACRO337882500 ACRO337880010
tert-Butyl (3R)-3-amino-3-phenylpropanoate 97%
tert-Butyl (3R)-3-amino-3-phenylpropanoate
Vzorec: C₁₃H₁₉NO₂
MW: 221,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00798309
CAS číslo: 161671-34-7
UN: 2735
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4960 to 1.4980 (20°C, 589 nm)
Specific optical rotation +17° to +22° (20°C, 589 nm) (c=1, CHCl3)
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR