Tisk…

Chloromethyl chlorosulphate 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Chloromethyl chlorosulphonate, Chlorosulphuric acid Chloromethyl Ester, Chlorosulphonic acid Chloromethyl Ester

330800010 330800050 330800250
ACRO330800010EA 2010 CZK
ACRO330800010 ACRO330800050 ACRO330800250
Chloromethyl chlorosulphate 97%
Chloromethyl chlorosulphonate
Vzorec: CH₂Cl₂O₃S
MW: 164,98 g/mol
Bod varu: 45…50 °C (10 mmHg)
Hustota: 1,6 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00041547
CAS číslo: 49715-04-0
UN: 3390
ADR: 6.1,

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4480 (20°C, 589 nm)
Titration Argentometric 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR