4-Amino-6-chlorobenzene-1,3-disulphonamide 98%
Dodavatel: Thermo Scientific
Synonyms:
4-Amino-6-chloro-1,3-benzenedisulphonamide
251850050
251851000
ACRO251850050EA
986
CZK
ACRO251850050
ACRO251851000
4-Amino-6-chlorobenzene-1,3-disulphonamide 98%
4-Amino-6-chlorobenzene-1,3-disulphonamide
Vzorec:
C₆H₈ClN₃O₄S₂ MW: 285,7300 g/mol Bod tání: 260…263 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007933 CAS číslo: 121-30-2 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 260 to 264 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...