Aflatoxin B1, crystalline
Dodavatel: Thermo Scientific
|
Nebezpečí
|
227340010
227340100
227340500
ACRO227340010EA
2160
CZK
ACRO227340010
ACRO227340100
ACRO227340500
Aflatoxin B1, crystalline
Aflatoxin B1
Vzorec:
C₁₇H₁₂O₆ MW: 312,3 g/mol Bod tání: 268…269 °C Storage Temperature: Freezer |
MDL Number:
MFCD00869647 CAS číslo: 1162-65-8 UN: 3462 ADR: 6.1,I |
Výsledky specifikačního testu
Appearance | White to yellow Powder |
Solubility | (10 mg/ml in CH2Cl2) Clear colorless to light yellow |
HPLC | ≥98 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...