Lactulose 99+%
Dodavatel: Thermo Scientific
225930250
225930050
225931000
ACRO225930250EA
5340
CZK
ACRO225930250
ACRO225930050
ACRO225931000
Lactulose 99+%
Lactulose
Vzorec:
C₁₂H₂₂O₁₁ MW: 342,297 g/mol Bod tání: 173…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063276 CAS číslo: 4618-18-2 |
Výsledky specifikačního testu
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (1 g, 105°C, 3 h) |
HPLC | ≥99.0 % (ELSD) |
Specific optical rotation | -46° to -50° (20°C, 589 nm) (c=1, H2O, Ammonia) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...