Tisk…

Chlorocarbonylsulphenyl chloride 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Chlorothioformyl Chloride, Chloroformylsulphenyl Chloride

224340050 224340250
ACRO224340050EA 2360 CZK
ACRO224340050 ACRO224340250
Chlorocarbonylsulphenyl chloride 95%
Chlorocarbonylsulphenyl chloride
Vzorec: CCl₂OS
MW: 130.97 g/mol
Bod varu: 98…100 °C (760 mmHg)
Hustota: 1.58 g/cm³
Bod vzplanutí: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000703
CAS číslo: 2757-23-5
UN: 3265
ADR: 8 II

Objednat

Výsledky specifikačního testu

Appearance Yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR