Tisk…

Bromoacetyl chloride 95%

Dodavatel: Thermo Scientific
Nebezpečí

197990250 197991000
ACRO197990250EA 2150 CZK
ACRO197990250 ACRO197991000
Bromoacetyl chloride 95%
Bromoacetyl chloride
Vzorec: BrCH₂COOCl
MW: 157,39 g/mol
Bod varu: 127…128 °C (760 mmHg)
Hustota: 1,9 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00000724
CAS číslo: 22118-09-8
UN: 3265
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4970 (20°C, 589 nm)
Titration Argentometric 94.0 to 106.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR