Palladium(II) oxide 99.995% (trace metals basis)
Dodavatel: Thermo Fisher Scientific
193680010
193680050
ACRO193680010EA
10590
CZK
ACRO193680010
ACRO193680050
Palladium(II) oxide 99.995% (trace metals basis)
Palladium(II) oxide
Vzorec:
OPd Hustota: 8,73 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011172 CAS číslo: 1314-08-5 |
Výsledky specifikačního testu
Appearance | Grey to black Powder or granules and/or chunks |
Assay | 85.2 to 88.7 % (Pd) |
Total trace metal impurities | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...