N,N-Dimethylformamide diethyl acetal 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1,1-Diethoxy-N,N-dimethylmethanamine, 1,1-Diethoxytrimethylamine
162080100
162080500
ACRO162080100EA
1230
CZK
ACRO162080100
ACRO162080500
N,N-Dimethylformamide diethyl acetal 95%
N,N-Dimethylformamide diethyl acetal
Vzorec:
C₇H₁₇NO₂ MW: 147.22 Bod varu: 128…130 °C (760 mmHg) Hustota: 0.859 g/cm³ Bod vzplanutí: 28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009227 CAS číslo: 1188-33-6 UN: 1993 ADR: 3 III |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3990 to 1.4010 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...