Tisk…
Safety ankle boots, lace-up, uvex quatro pro, 8401
Dodavatel: UVEX
8401.2/39 8401.2/40 8401.2/41 8401.2/42 8401.2/43 8401.2/44 8401.2/45 8401.2/46 8401.2/47 8401.2/48 8401.2/49 8401.2/50
114-7062EA 3380 CZK
114-7062 114-7063 114-7064 114-7065 114-7066 114-7067 114-7068 114-7069 114-7070 114-7071 114-7072 114-7073
Safety ankle boots, lace-up, uvex quatro pro, 8401
Obuv
Sturdy and comfortable anti-static safety ankle boot made from water resistant leather upper, soft padded collar and padded dust tongue and a dual-density, self-cleaning and slip-resistant PUR sole (4,5 mm, heat resistant to +120 °C, oil and petrol resistant, silicone-free).
  • Climazone technology for improved comfort
  • Breathable textile lining
  • Removable, anti-static insole with a moisture control system creates a superior foot climate and improved comfort
  • uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Sole offers integrated heel support
  • Smooth-running, dirt-resistant lacing system
  • Foamed scuffcap for additional protection
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certifikace: DGUV 112-191; EN ISO 20345:2011; S3, SRC