Tisk…

L-(+)-Tartaric acid dipotassium salt 97%

Dodavatel: Thermo Scientific

Synonyms: L-2,3-Dihydroxybutanedioic acid dipotassium salt

427991000 427995000
ACRO427991000EA 1370 CZK
ACRO427991000 ACRO427995000
L-(+)-Tartaric acid dipotassium salt 97%
L-(+)-Tartaric acid dipotassium salt
Vzorec: C₄H₄K₂O₆
MW: 226,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013065
CAS číslo: 921-53-9

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤4 % (150°C, 4 hr)
Specific optical rotation +25° to +27° (20°C, 589 nm) (c=1,H2O)
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR