Isobutylmagnesium bromide 2 M in diethyl ether, AcroSeal®
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
427391000
427398000
427391000.EA
4450
CZK
427391000.
427398000.
Isobutylmagnesium bromide 2 M in diethyl ether, AcroSeal®
Isobutylmagnesium bromide
Appearance: Yellow to dark brown Solution
Vzorec:
C₄H₉BrMg Bod varu: 34,6 °C (760 mmHg) Hustota: 0,941 g/cm³ Bod vzplanutí: –34 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075126 CAS číslo: 926-62-5 UN: 3399 ADR: 4.3,I |
Výsledky specifikačního testu
Appearance | Yellow to dark brown Solution |
Titration Grignard | 1.9 to 2.2 M |
Specific surface area | 35 to 65 m²/g |
pH | 3.5 to 4.5 (4 % dispersion) |
Acidity (HCl) | ≤0.3 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...