3-Oxocyclopentanecarboxylic acid 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-oxocyclopentane-1-carboxylic acid, 3-Oxo-1-cyclopentanecarboxylic acid, 3-Oxo-cyclopentane-carboxylic acid
426340010
426340050
ACRO426340010EA
5370
CZK
ACRO426340010
ACRO426340050
3-Oxocyclopentanecarboxylic acid 97%
3-Oxocyclopentanecarboxylic acid
Appearance: White to yellow Crystalline powder
Vzorec:
C6H8O3 Storage Temperature: Ambient |
MDL Number:
MFCD01320173 CAS číslo: 98-78-2 |
Výsledky specifikačního testu
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 59 to 64 °C |
NMR (1H-NMR) | Consistent with structure |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...