Tisk…

Dimethyl chlorophosphate 96%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Chlorophosphoric acid dimethyl ester, Dimethyl phosphorochloridate

426140050 426140250
ACRO426140050EA 2850 CZK
ACRO426140050 ACRO426140250
Dimethyl chlorophosphate 96%
Dimethyl chlorophosphate
Vzorec: C₂H₆ClO₃P
MW: 144,49 g/mol
Bod varu: 80 °C (25 mmHg)
Bod vzplanutí: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00117904
CAS číslo: 813-77-4
UN: 2927
ADR: 6.1,I

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4140 (20°C, 589 nm)
Titration Argentometric ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR