Tisk…

5-Carboxyfluorescein 95%

Dodavatel: Thermo Fisher Scientific
Varování

425811000 425815000
ACRO425811000EA 3260 CZK
ACRO425811000 ACRO425815000
5-Carboxyfluorescein 95%
5-Carboxyfluorescein

Appearance: Yellow to orange Powder

Vzorec: C₂₁H₁₂O₇.xH₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00036874
CAS číslo: 76823-03-5

Objednat

Výsledky specifikačního testu

Appearance Yellow to orange Powder
Infrared spectrum Conforms
HPLC ≥94 %
NMR (1H-NMR) Consistent with structure
Sulfated ash ≤0.1 %
Solubility (in water) Sparingly soluble
Related substances Passes test
Titration with NaOH 99.5 to 100.5 % (On dried substance)
Chloride (Cl) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR