2-(3-Butynyloxy)tetrahydro-2H-pyran 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(But-3-yn-1-yloxy)tetrahydro-2H-pyran
428210050
428210250
ACRO428210050EA
2780
CZK
ACRO428210050
ACRO428210250
2-(3-Butynyloxy)tetrahydro-2H-pyran 97%
2-(3-Butynyloxy)tetrahydro-2H-pyran
Appearance: Clear colorless to light yellow Liquid
Vzorec:
C₉H₁₄O₂ MW: 154,209 g/mol Bod varu: 92…95 °C (18 mmHg) Hustota: 0,984 g/cm³ Bod vzplanutí: 73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012352 CAS číslo: 40365-61-5 |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
NMR | consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...