Tisk…

Mithramycin A (from Streptomyces argillaceus) 90%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

415090050 415090010
ACRO415090050EA 16660 CZK
ACRO415090050 ACRO415090010
Mithramycin A (from Streptomyces argillaceus) 90%
Mithramycin A

Appearance: Yellow Powder


Soluble in DMSO (100 mg/mL)

Vzorec: C₅₂H₇₆O₂₄
Bod tání: 180…183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135618
CAS číslo: 18378-89-7
UN: 2811
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance Yellow Powder
Solubility (100 mg/ml in DMSO) Clear yellow
HPLC ≥88 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR