Tisk…

(Dimethylamino)acetaldehyde dimethylacetal 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2,2-Dimethoxy-N,N-dimethylethanamine, N-(2,2-Dimethoxyethyl)dimethylamine

390910010 390910050
ACRO390910010EA 967 CZK
ACRO390910010 ACRO390910050
(Dimethylamino)acetaldehyde dimethylacetal 97%
(Dimethylamino)acetaldehyde dimethylacetal
Vzorec: C₆H₁₅NO₂
Bod varu: 64…66 °C (12 mmHg)
Bod vzplanutí: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00010205
CAS číslo: 38711-20-5
UN: 2920
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4120 (20°C, 589 nm)
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR