Tisk…

Magnesium tert-butoxide 93+%

Dodavatel: Thermo Fisher Scientific
Varování

390470100 390470500 390472500
ACRO390470100EA 1310 CZK
ACRO390470100 ACRO390470500 ACRO390472500
Magnesium tert-butoxide 93+%
Magnesium tert-butoxide

Appearance: White to light grey Powder

Vzorec: C₈H₁₈MgO₂
MW: 170.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788761
CAS číslo: 32149-57-8
UN: 3205
ADR: 4.2 II

Objednat

Výsledky specifikačního testu

Appearance White to light grey Powder
Titration with HCl ≥93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR