Potassium vinyltrifluoroborate 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
387700010
387700050
ACRO387700010EA
1600
CZK
ACRO387700010
ACRO387700050
Potassium vinyltrifluoroborate 95%
Potassium vinyltrifluoroborate
Vzorec:
C₂H₃BF₃K Bod tání: 240…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093335 CAS číslo: 13682-77-4 UN: 3261 ADR: 8,II |
Výsledky specifikačního testu
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
Assay | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...