3-bromofuran-2,5-dione 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Bromomaleic anhydride
147450100
147450500
ACRO147450100EA
2340
CZK
ACRO147450100
ACRO147450500
3-bromofuran-2,5-dione 97%
3-bromofuran-2,5-dione
Appearance: Clear colorless to yellow Liquid
Vzorec:
C₄HBrO₃ MW: 176,953 g/mol Bod varu: 215 °C (760 mmHg) Hustota: 1,905 g/cm³ Bod vzplanutí: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005519 CAS číslo: 5926-51-2 UN: 3265 ADR: 8,III |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5410 to 1.5430 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...