Tisk…
N,N-Diisopropylethylamine trihydrofluoride 90%
Dodavatel: Thermo Fisher Scientific
Nebezpečí
Synonyms: N-Ethyl-N-isopropylpropan-2-amine trihydrofluoride
390710050 390710010
ACRO390710050EA 5080 CZK
ACRO390710050 ACRO390710010
N,N-Diisopropylethylamine trihydrofluoride 90%
N,N-Diisopropylethylamine trihydrofluoride
Appearance: Clear colorless to yellow-brown Liquid
Vzorec: C₈H₂₂F₃N
Bod varu: 93…95 °C (4 mmHg)
Hustota: 0,965 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00144880
CAS číslo: 131600-43-6
UN: 3286
ADR: 3,II
Objednat
Výsledky specifikačního testu
Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Titration with NaOH 18 to 22 % (available HF)