Tisk…

Isonicotinoyl chloride hydrochloride 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Pyridine-4-carbonyl Chloride Hydrochloride, 4-Pyridinoyl chloride hydrochloride

211680250 211681000 211682500
ACRO211680250EA 2720 CZK
ACRO211680250 ACRO211681000 ACRO211682500
Isonicotinoyl chloride hydrochloride 97%
Isonicotinoyl chloride hydrochloride

Appearance: White to beige Crystalline powder

Vzorec: C₆H₄ClNO
MW: 178,01 g/mol
Bod tání: 153…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00012830
CAS číslo: 39178-35-3
UN: 3261
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 153 to 161 °C
Titration Argentometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR