Tisk…

Benzoylnitromethane 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Nitro-1-phenylethanone

372790050 372790250
ACRO372790050EA 2430 CZK
ACRO372790050 ACRO372790250
Benzoylnitromethane 98%
Benzoylnitromethane

Appearance: White to yellow to tan Powder or flakes

Vzorec: C₈H₇NO₃
Bod tání: 101…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010218
CAS číslo: 614-21-1

Objednat

Výsledky specifikačního testu

Appearance White to yellow to tan Powder or flakes
Infrared spectrum Conforms
Melting point 102 to 107 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR